50 X 25 TUBETESTS COD LR (0 - 150 MG/L) 1 * 1.250 Vial
Catalog # WTIDCOD-79-LR-50
Supplier: WATER ID PRIMELAB
50 X 25 TUBETESTS COD LR (0 - 150 MG/L) 1 * 1.250 Vial
Catalog # WTIDCOD-79-LR-50
Supplier: WATER ID PRIMELAB
Frequently Bought Together
Visual colour kits, ammonium, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide, Supelco®
Supplier: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
Test kits, cyanide, Spectroquant®, Supelco®
Supplier: Merck
Formic acid ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Test strips for copper, MQuant®, Supelco®
Supplier: Merck
Sodium nitrite 0.1 N, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
Supplier: Merck
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Test strips for peroxide, MQuant®, Supelco®
Supplier: Merck
Sodium hydroxide 32% in aqueous solution, GPR RECTAPUR®, pure for nitrogen analysis
Supplier: VWR Chemicals
Test kits, TOC, Spectroquant®, Supelco®
Supplier: Merck
Test strips for formaldehyde, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, Nitrite, LCK cuvette tests
Supplier: Hach
Test kits, photometric, iron, LCK cuvette tests
Supplier: Hach
Test kits, photometric, Zirconium, LCK cuvette tests
Supplier: Hach