Order Entry
Spain
ContactUsLinkComponent
22 components: Ag 10mg/l Al 10mg/l B 10mg/l Ba 10mg/l Ca 10mg/l Cd 10mg/l Co 10mg/l Cr 10mg/l Cu 10mg/l Sn 10mg/l Fe 10mg/l Mg 10mg/l Mn 10mg/l Mo 10mg/l Ni 10mg/l Pb 10mg/l K 5000mg/l Na 10mg/l S 100mg/l Sr 10mg 1 * 100 mL
Catalog # CMPLRM092643L1
22 components: Ag 10mg/l Al 10mg/l B 10mg/l Ba 10mg/l Ca 10mg/l Cd 10mg/l Co 10mg/l Cr 10mg/l Cu 10mg/l Sn 10mg/l Fe 10mg/l Mg 10mg/l Mn 10mg/l Mo 10mg/l Ni 10mg/l Pb 10mg/l K 5000mg/l Na 10mg/l S 100mg/l Sr 10mg 1 * 100 mL
Catalog # CMPLRM092643L1