XSelect HSS Cyano (CN) Method Validation Kit, 100Å, 5 µm, 4.6 mm X 100 mm 1 * 3 items
Catalog # WATE186005958
Supplier: WATERS
XSelect HSS Cyano (CN) Method Validation Kit, 100Å, 5 µm, 4.6 mm X 100 mm 1 * 3 items
Catalog # WATE186005958
Supplier: WATERS
Frequently Bought Together
VWR®, Manual Crimper and Decapper
Supplier: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Vial racks for headspace vials
Supplier: Restek
VWR®, Screw Caps for Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Neck Vials ND8
Supplier: VWR Collection
VWR®, Septa GL 45
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND24
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR® Air Valve
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Screw Neck Vials ND24
Supplier: VWR Collection