ACQUITY UPLC HSS Cyano (CN) Column, 100Å, 1.8 µm, 2.1 mm X 100 mm 1 * 3 items
Catalog # WATE176002709
Supplier: WATERS
ACQUITY UPLC HSS Cyano (CN) Column, 100Å, 1.8 µm, 2.1 mm X 100 mm 1 * 3 items
Catalog # WATE176002709
Supplier: WATERS
Frequently Bought Together
Total organic carbon (TOC) vials
Supplier: EP SCIENTIFIC
VWR®, Screw Neck Headspace Vials ND18
Supplier: VWR Collection
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Screw Caps for Screw Neck Vials ND13
Supplier: VWR Collection
Air valves, b.safe
Supplier: Bohlender
EPA screw neck vials, N 24
Supplier: MACHEREY-NAGEL
Glass capillaries and graduated micropipettes, disposable
Supplier: CAMAG
Avantor® ACE® Phenyl, HPLC Columns, 3 µm
Supplier: Avantor
Avantor® ACE® C18, HPLC Columns, 5 µm
Supplier: Avantor
Chromatography Tubing, Stainless Steel
Supplier: Avantor
Supplies for pumps
Supplier: Agilent
SafetyWasteCaps
Supplier: S.C.A.T.
QuEChERS mixes, clean-up, CHROMABOND®
Supplier: MACHEREY-NAGEL
Accessories for sprayer
Supplier: LENZ, E. (Lenz Laborglas GmbH & CO.KG)
VWR®, Crimp Neck Headspace Vials ND20
Supplier: VWR Collection