CONTAINMENT SEAL FOR PRECELLYS 24 TOUCH 1 * 1 items
Catalog # BERTP002535P24T0A0
Supplier: Bertin Instruments
CONTAINMENT SEAL FOR PRECELLYS 24 TOUCH 1 * 1 items
Catalog # BERTP002535P24T0A0
Supplier: Bertin Instruments
Frequently Bought Together
Ethanol, GPR RECTAPUR®
Supplier: VWR Chemicals
Gas cartridges, Campingaz®
Supplier: CAMPING GAZ
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Hypodermic needles, HENKE-JECT® for single use
Supplier: HENKE SASS WOLF
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Accessories for accu-jet® pro
Supplier: Brand
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals
Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck