K 0.5MG/L IN HNO3 2% 1 * 100 mL
Catalog # CMPLRM091058L1
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Supplier: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Supplier: Merck
NANOCOLOR® Photometric Tube Test Kits for Phosphates (Ortho) and Phosphates (Total)
Supplier: MACHEREY-NAGEL
Immersion oil for microscopy, Sigma-Aldrich®
Supplier: Merck
Trifluoroacetic acid ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Visual colour kits, chlorine, MQuant®, Supelco®
Supplier: Merck
Test strips, peracetic acid, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Sodium hypochlorite solution 5%, J.T.Baker®
Supplier: Avantor
Hydrochloric acid 2 mol/l (2 N)
Supplier: PROLABO CMP
Test strips, peroxide, QUANTOFIX®
Supplier: MACHEREY-NAGEL
Test kits, chlorine dioxide, Spectroquant®, Supelco®
Supplier: Merck
Test strips for free chlorine, MQuant®, Supelco®
Supplier: Merck
Test kits, photometric, Nitrite, LCK cuvette tests
Supplier: Hach
Standards, calibration for LCK cuvettes, Addista
Supplier: Hach
Test strips
Supplier: Hach