3,5-DICHLOROBENZENE-1-CARBOXIMIDAMIDE HY 1 * 1 g
Catalog # APOSOR29563-1G
Supplier: Apollo Scientific
3,5-DICHLOROBENZENE-1-CARBOXIMIDAMIDE HY 1 * 1 g
Catalog # APOSOR29563-1G
Supplier: Apollo Scientific
Frequently Bought Together
Formamide, EMSURE® for analysis, Supelco®
Supplier: Merck
1,3-Dioxolane ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Supplier: VWR Chemicals
Methanol, anhydrous (max. 0.002% H₂O) ≥99.9%
Supplier: VWR Chemicals
Dimethyl sulphoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: Avantor
2-Propanol 99+%, meets analytical specification of Ph.Eur.
Supplier: Thermo Fisher Scientific
Acetone ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
N-Methyl-2-pyrrolidone (NMP), anhydrous ≥99.5%
Supplier: Thermo Fisher Scientific
Ethylenediamine ≥99.0% (by GC), GPR RECTAPUR®
Supplier: VWR Chemicals
2-Ethoxyethanol 99%, Reagent Grade
Supplier: Honeywell Chemicals
Ethanol absolute, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor