ADENOVIRUS ANTIBODY 8C4 [ALEXA FLUOR 488 1 * 0,2 mL
NOVUNB600-413AF488
Frequently Bought Together
Roller bottle standard, surface treated, 1000 ml vent cap 1 * 24 items
: GUANGZHOU JET BIOFILTRATION
MULTIP. BEAKER PP 120 ML ST 1 * 300 items
: Greiner Bio-One
PLATE COUNT ASEP 90/20 1 * 1 KIT
: DIFCO BD
24 Well, OPEN Reaction Block for 15mm O 1 * 1 items
: MARKET SOURCE PART PROCESS
Tubes Cryo.S™ 4 and 5 ml
: Greiner Bio-One
Multidishes, Nunc™ UpCell™ surface
: Thermo Fisher Scientific
VWR®, Cell Culture Insert Plates, PET
: VWR Collection
Culture tubes, sterile, Cultubes™
: Simport Scientific
VWR® Square Polycarbonate Media Bottles
: VWR Collection
384-WELL COLLECTION PLATE, 1-90L WORKING VOLUME 1 * 100 UN
: MARKET SOURCE PART PROCESS
Sample containers with screw cap
: DELTALAB