Formaldehyde solution Emprove Evolve 1 * 950 L
Catalog # 1.04002.9950
Supplier: MERCK PRODUCTION CHEMICALS
Formaldehyde solution Emprove Evolve 1 * 950 L
Catalog # 1.04002.9950
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Chloroform ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Acetic anhydride for synthesis, Sigma-Aldrich®
Supplier: Merck
Glycerine 87% 86%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
Glycerine ≥98%, GPR RECTAPUR®
Supplier: VWR Chemicals
4-Aminobenzoic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Iodomethane ≥99% stabilised
Supplier: Thermo Fisher Scientific
Dichloromethane ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 60…80 °C
Supplier: Thermo Fisher Scientific
N,N-Dimethylformamide, extra dry over Molecular Sieve 99.8%, AcroSeal®
Supplier: Thermo Fisher Scientific
m-Xylene ≥99.0% (by GC)
Supplier: TCI
1-Propanol ACS
Supplier: Thermo Fisher Scientific
Tetrahydrofuran ≥99% stabilised
Supplier: Thermo Fisher Scientific