Additional shelves for FD Range Ovens
Stainless steel shelf, for heavy loads, max load 70 kg, Reels BF/FD/FED 720
Catalog # BIND8012-2185
Supplier: Binder
Jump to:
Product Details & Documents
Specifications
- Description:Stainless steel shelf, for heavy loads, max load 70 kg
- Seq:05
- For:Reels BF/FD/FED 720