Sodium sulfate anhydrous EMPROVE EXPERT Ph Eur,BP,ChP,USP 1 * 3 items
1.28225.0003
Sodium sulfate anhydrous EMPROVE EXPERT Ph Eur,BP,ChP,USP 1 * 3 items
1.28225.0003
Frequently Bought Together
Sodium hydrogen carbonate ≥98%, TECHNICAL
: VWR Chemicals
Potassium iodide ≥99%, GPR RECTAPUR®
: VWR Chemicals
Calcium carbonate ≥99%, GPR RECTAPUR®, precipitated
: VWR Chemicals
Iodine ≥99%, GPR RECTAPUR®, resublimed
: VWR Chemicals
Sodium hypochlorite 4.00 - 4.99% available chlorine in aqueous solution, Reagent Grade
: Honeywell Chemicals
Sodium acetate, anhydrous 99%
: Thermo Fisher Scientific
Sodium hypochlorite (11 - 14% Cl₂) in aqueous solution
: Thermo Fisher Scientific