Order Entry
Spain
ContactUsLinkComponent
Detect CRE-colonized patients, a chromogenic screening plate that provides results in just 18 hours for carbapenem-resistant Enterobacteriaceae (CRE) 1 * 10 items
Catalog # OXOIPO1226A
Detect CRE-colonized patients, a chromogenic screening plate that provides results in just 18 hours for carbapenem-resistant Enterobacteriaceae (CRE) 1 * 10 items
Catalog # OXOIPO1226A
Frequently Bought Together