1,2-DICHLOROBENZENE 1 * 100 mg
Catalog # 36707-100MG
Supplier: Merck
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions, NIST traceable, Orion™
Supplier: Thermo Orion
Tin(II) chloride 30% in hydrochloric acid 20% Reag. Ph. Eur. 1085001
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
Lithium standard for IC, TraceCERT®, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
Conductivity standards, KCl, referenced at 25 °C, for conductivity measurement
Supplier: Mettler - Toledo
Buffer solutions in sachets
Supplier: Mettler - Toledo
pH Standard Solutions, IUPAC, Certified
Supplier: Hach