2-(METHACRYLOYLOXY)-N,N,N-TRIMETHYLETHA 1 * 25 g
Catalog # TCIAM3306-25G
Supplier: TCI
Frequently Bought Together
Cyclohexanone ≥99.0% analytical reagent
Supplier: VWR Chemicals
Petroleum spirit, 80…110 °C, TECHNICAL
Supplier: VWR Chemicals
Dichloromethane ≥99.5% stabilised, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Benzoic acid methyl ester for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
tert-Butyl methyl ether anhydrous, 99.8 % 1 * 1 L
Supplier: Merck
Xylene (mixture of isomers) ≥98%, liquid
Supplier: PanReac AppliChem
Acetone ≥99.5%, AR®, Macron Fine Chemicals™
Supplier: MACRON AVANTOR BRAND
2-Propanol ≥99.8% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Methanol 99.9%, gradient grade for HPLC
Supplier: Thermo Fisher Scientific
Dichloromethane, anhydrous ≥99.7% stabilised
Supplier: Thermo Fisher Scientific
N,N-Dimethylacetamide ≥99.5% (by GC), Puriss. p.a., Riedel-de Haën™
Supplier: Honeywell Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
Supplier: Honeywell Chemicals
Diethyl ether stabilised, BAKER, J.T.Baker®
Supplier: Avantor