Order Entry
Spain
Orders LinkContactUsLinkComponent
Multicollector, b.safe
Multicollector, b.safe
Multicollector, 4× GL 45 (male), GL 45 (female)
Catalog # BOHLR1140-04
Supplier:   Bohlender
Multicollector, b.safe
Catalog # BOHLR1140-04
Supplier:  Bohlender
Multicollector, b.safe
Multicollector, b.safe
Multicollector, 4× GL 45 (male), GL 45 (female)
Price
208,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Combination of capillaries and tubes of several HPLC systems in one desktop mount.
  • With a b.safe Multicollector you simply multiply the number of connectors of a b.safe Desktop Mount
  • This extension can be done without any modification of the existing installation
  • Chemically inert
  • Static dissipative to prevent static charging
Made of static dissipative PE-EX, screw cap made of PPS with GL 45 thread, rotary insert of static dissipative PE-EX and male threads GL 45. To connect up to 4 b.safe System Caps GL 45 to one desktop mount GL 45 (not included in scope of delivery).
Specifications
  • Thread:
    4× GL 45 (male), GL 45 (female)
  • Description:
    Multicollector
  • Seq:
    02