Multicollector, b.safe
Multicollector, 3× GL 45 (male), GL 45 (female)
Catalog # BOHLR1140-03
Supplier: Bohlender
Jump to:
Product Details & Documents
Combination of capillaries and tubes of several HPLC systems in one desktop mount.
- With a b.safe Multicollector you simply multiply the number of connectors of a b.safe Desktop Mount
- This extension can be done without any modification of the existing installation
- Chemically inert
- Static dissipative to prevent static charging
Made of static dissipative PE-EX, screw cap made of PPS with GL 45 thread, rotary insert of static dissipative PE-EX and male threads GL 45. To connect up to 4 b.safe System Caps GL 45 to one desktop mount GL 45 (not included in scope of delivery).
Documents
Cutsheet BOHLR1140-03 R 1140-03 Cutsheet BOHLR1140-04 R 1140-04
Technical Drawing
Specifications
- Thread:3× GL 45 (male), GL 45 (female)
- Description:Multicollector
- Seq:01