Order Entry
Spain
ContactUsLinkComponent
Validated qPCR-kit for rapid fungal contamination control of ATMPs containing 100 reactions store at 2-8 °C in the dark 1 * 1 KIT
Catalog # SARTSMB95-1012
Supplier:  Sartorius
Validated qPCR-kit for rapid fungal contamination control of ATMPs containing 100 reactions store at 2-8 °C in the dark 1 * 1 KIT
Catalog # SARTSMB95-1012
Supplier:  Sartorius
Frequently Bought Together