Order Entry
Spain
Orders LinkContactUsLinkComponent
 

γ-Cyclodextrine 98%

Bottle, 25 g
Catalog # APOSBIB6136-25G
Irritant
Supplier:   Apollo Scientific
γ-Cyclodextrine 98%
Catalog # APOSBIB6136-25G
Supplier:  Apollo Scientific
CAS Number:  17465-86-0
γ-Cyclodextrine 98%

Bottle, 25 g

CAS Number:  17465-86-0
Price
46,40 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  gamma-Cyclodextrin, Cyclooctaamylose, γ-Cyclodextrin, Schardinger gamma-Dextrin
  • CAS Number:17465-86-0
  • Formula:C₄₈H₈₀O₄₀
  • MDL Number:MFCD00009595
  • Molecular Weight:1297.1248 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.