Human CD62E ELISA Set (without plates)
Human CD62E ELISA Set (without plates), 10×96 tests
Catalog # ABCAAB47348-10X96
Supplier: Abcam
Jump to:
Product Details & Documents
Human CD62E ELISA Set (without plates) is a Sandwich (quantitative) ELISA kit for the measurement of Human CD62E ELISA Set (without plates) in Human in Plasma, Cell culture supernatant, Serum samples.
Specifications
Specifications
- Description:Human CD62E ELISA Set (without plates)
- Tests per kit:10×96 tests
- Primary antibody reactivity:Human
- Sample type:Plasma, Cell culture supernatant, Serum
- Detection range:1 - 32 ng/ml
- Storage temperature:4 °C
- Assay Type:Sandwich
- Time to Results:2 h 15 min
- Cat. No.:ABCAAB47348-10X96
- Assay duration:Multiple steps
- Detection method:Colorimetric
- Sensitivity:0,5 ng/ml
- Format:Pre-coated
- Target protein:CD62E
- Regulatory status:RUO
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



