PIPERACILLIN FOR PEAK IDENTIFICATION 1 * 15 mg
Catalog # EDQMY0002235
Supplier: EDQM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Removable needles
Supplier: HAMILTON BONADUZ
Citric acid monohydrate
Supplier: EDQM