Entrada de pedido
Spain
Orders LinkContactUsLinkComponent
b.safe Funnels 130
b.safe Funnels 130
Static dissipative funnel with grounding cable, S60
Catalog # 590-0383
Supplier:   Bohlender
b.safe Funnels 130
Catalog # 590-0383
Supplier:  Bohlender
b.safe Funnels 130
Static dissipative funnel with grounding cable, S60
Price
105,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Funnel made of PE or static dissipative PE-EX for liquid waste disposal. The funnel can be safely mounted on the waste container with its freely turnable cap that is safely fixed with filling neck.
  • Smooth-running hinged lid protects from harmful vapours
  • Removable sieve restrains e.g. magnetic stirring bars or other solids
  • Small installation height ideal for integration into safety cabinets
  • All static dissipative funnels are supplied with assembled grounding cable (lengh: 1,5 m)
  • Free turnable cap for easy assembly on collecting container
  • Neck with plastic nibs to fix attachable fitting tubes for under level filling
The sieve restrains magnetic stirrer bars and other solid materials. It has a grip for easy removal for cleaning. The hinged lid reliably withholds vapours from the container.
The static dissipative PE-EX version includes two attachable filling tubes (Ø 28 × usable length 97 mm per tube). The single segments are permanently fixed by two clip-in nibs and can be shortened with an appropriate saw. Safe earthing of the funnel can be made via the 1,5 m long grounding cable by qualified personnel. The mounted filling tube allows an under level filling to avoid static charging.
Documents
Specifications
  • Rosca:
    S60
  • Descripción:
    Static dissipative funnel with grounding cable
  • Seq:
    11
Accessories
Product Information
  • Catalog #
    Descripción
    Availability
    Price
  • 590-0313
    Static dissipative sieve for funnel 130
    Log in for availability
    1 * 1 UN - 15,50 €
Product Information
  • Catalog #
    Descripción
    Availability
    Price
  • 590-0314
    Sieve for funnel 130
    Log in for availability
    1 * 1 UN - 24,20 €
Product Information
  • Catalog #
    Descripción
    Availability
    Price
  • 590-0317
    Static dissipative filling tubes for funnel 130
    Log in for availability
    1 * 5 UN - 52,10 €
Product Information
  • Catalog #
    Descripción
    Availability
    Price
  • 590-0318
    Filling tubes for funnel 130
    Log in for availability
    1 * 5 UN - 16,30 €
Product Information
  • Catalog #
    Descripción
    Availability
    Price
  • 590-0333
    Static dissipative angled adaptor for canisters, for vertical adjustment of b.safe Funnels 130 with thread S60 on b.safe canisters S60 with inclined neck
    Log in for availability
    1 * 1 UN - 114,00 €