Order Entry
Spain
Orders LinkContactUsLinkComponent
b.safe Funnels 130
b.safe Funnels 130
Funnel, S55
Catalog # 590-0378
Supplier:   Bohlender
b.safe Funnels 130
Catalog # 590-0378
Supplier:  Bohlender
b.safe Funnels 130
Funnel, S55
Price
88,30 €
Log in for availability
1 * 1 UN
Product Details & Documents
Funnel made of PE or static dissipative PE-EX for liquid waste disposal. The funnel can be safely mounted on the waste container with its freely turnable cap that is safely fixed with filling neck.
  • Smooth-running hinged lid protects from harmful vapours
  • Removable sieve restrains e.g. magnetic stirring bars or other solids
  • Small installation height ideal for integration into safety cabinets
  • All static dissipative funnels are supplied with assembled grounding cable (lengh: 1,5 m)
  • Free turnable cap for easy assembly on collecting container
  • Neck with plastic nibs to fix attachable fitting tubes for under level filling
The sieve restrains magnetic stirrer bars and other solid materials. It has a grip for easy removal for cleaning. The hinged lid reliably withholds vapours from the container.
The static dissipative PE-EX version includes two attachable filling tubes (Ø 28 × usable length 97 mm per tube). The single segments are permanently fixed by two clip-in nibs and can be shortened with an appropriate saw. Safe earthing of the funnel can be made via the 1,5 m long grounding cable by qualified personnel. The mounted filling tube allows an under level filling to avoid static charging.
Documents
Specifications
  • Thread:
    S55
  • Description:
    Funnel
  • Seq:
    04
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Static dissipative sieve for funnel 130
    Static dissipative sieve for funnel 130
    590-0313
    Log in for availability
    1 * 1 UN - 15,50 €
Product Information
  • Catalog #
    Availability
    Price
  • Sieve for funnel 130
    Sieve for funnel 130
    590-0314
    Log in for availability
    1 * 1 UN - 24,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Static dissipative filling tubes for funnel 130
    Static dissipative filling tubes for funnel 130
    590-0317
    Log in for availability
    1 * 5 UN - 52,10 €
Product Information
  • Catalog #
    Availability
    Price
  • Filling tubes for funnel 130
    Filling tubes for funnel 130
    590-0318
    Log in for availability
    1 * 5 UN - 16,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Static dissipative angled adaptor for canisters, for vertical adjustment of b.safe Funnels 130 with thread S60 on b.safe canisters S60 with inclined neck
    Static dissipative angled adaptor for canisters, for vertical adjustment of b.safe Funnels 130 with thread S60 on b.safe canisters S60 with inclined neck
    590-0333
    Log in for availability
    1 * 1 UN - 114,00 €