Flat seals
Flat seal, fits outer thread, 50,8 mm, PE
Catalog # BURK8538-0200
Supplier: BURKLE
Jump to:
Product Details & Documents
These flat seals are made from transparent PE.
- Fits outer or inner thread
Specifications
- Description:Flat seal, fits outer thread, 50,8 mm
- Material:PE