Microplate readers, Multiskan SkyHigh
Product Details & Documents
The Multiskan SkyHigh is a UV/Visible microplate spectrophotometer designed to be convenient and easy to use for virtually any photometric research application, especially DNA, RNA and protein analysis. It is ideal for multi-user environments where a variety of endpoint, kinetic and spectral assays are performed.
- Fast, easy measurement - low volume nucleic acid or protein samples
- Simple to use software - enables experiment set-up within minutes
- Remote access - manage your data from anywhere via the Cloud, USB, or the Microsoft™ OneDrive™ platform
Multiskan Sky touch screen models can be connected to Thermo Fisher Connect cloud-based tools to enable secure digital data management, remote access of your instrument’s photometric data, and sharing between colleagues.
SkanIt™ software offers a comprehensive selection of built-in calculations, including blank subtraction, standard curves and concentration/dose-response curve fitting, classification, quality control, kinetic, and spectral analysis. Each user can install SkanIt software on his/her own computer to set up assays and analyse data. Thermo Fisher Connect cloud-based capabilities enable you to:
- Monitor the status of your instrument from your PC or Mac® computer
- Monitor the status of your instrument from your mobile device with the Instrument Connect app
- Upload measured data automatically or manually from your instrument to your cloud account
- Securely store, access, share, and manage your data remotely
- Receive automatic notification of new instrument software updates
- Upgrade instrument software at your convenience
Specifications
- Description:Multiskan SkyHigh with touch screen and cuvette
- Incubation range:From ambient temperature to 45 ˚C
- Light source:Xenon flash lamp
- Shaking:Linear
- Measurement time (96-well plate):6 s
- Measurement time (384-well plate):10 s
- Range:Up to 4 Abs
- Scan speed:10 s from 200 - 1000 nm in 1 nm steps
- Weight:11,3 kg
- Interfaces:1× USB B port for PC
1× Ethernet port
3× USB A ports for devices (USB memory device and Wi-Fi dongle) - Cat. No.:LABLA51119700C
- Accuracy:1,0% + 0,003 Abs (0 - 2,0 Abs), 2,0% (2,0 - 2,5 Abs)
- Wavelength selection:Monochromator
- Precision:SD <0,003 Abs or CV <1,0% (at 450 nm)
- User interface:Stand alone use: 7" touch screen display; PC control: Thermo Scientific™ SkanIt™ software
- Linearity:0 - 2,5 Abs, 2% (at 450 nm)
- Bandwidth:<2,5 nm
- Wavelength range:200 - 1000 nm (1 nm steps)
- W×D×H:295×445×265 mm
- Plate types:6- to 48-, 96-, and 384-well plates (max. height 19,5 mm with lid), Thermo Scientific™ µDrop™ Plates
- Cuvette type:W×D×H: 12,5×12,5×40 - 58 mm
Beam centre height: 8,5 mm
Beam window: ≥2
Accessories
Product Information
- Catalog #AvailabilityPrice
- µDrop™ Plate, 16 measurement positions in a 2×8 matrixµDrop™ Plate, 16 measurement positions in a 2×8 matrix736-0754Log in for availability1 * 1 UN - 3 290,00 €
Product Information
- Catalog #AvailabilityPrice
- µDrop™ Duo plate, 2× 16 measurement positions in two 2×8 matrixesµDrop™ Duo plate, 2× 16 measurement positions in two 2×8 matrixesLABLN12391M2Log in for availability1 * 1 UN - 3 570,00 €



