Microplate readers, Multiskan SkyHigh
Product Details & Documents
The Multiskan SkyHigh is a UV/Visible microplate spectrophotometer designed to be convenient and easy to use for virtually any photometric research application, especially DNA, RNA and protein analysis. It is ideal for multi-user environments where a variety of endpoint, kinetic and spectral assays are performed.
- Fast, easy measurement - low volume nucleic acid or protein samples
- Simple to use software - enables experiment set-up within minutes
- Remote access - manage your data from anywhere via the Cloud, USB, or the Microsoft™ OneDrive™ platform
Multiskan Sky touch screen models can be connected to Thermo Fisher Connect cloud-based tools to enable secure digital data management, remote access of your instrument’s photometric data, and sharing between colleagues.
SkanIt™ software offers a comprehensive selection of built-in calculations, including blank subtraction, standard curves and concentration/dose-response curve fitting, classification, quality control, kinetic, and spectral analysis. Each user can install SkanIt software on his/her own computer to set up assays and analyse data. Thermo Fisher Connect cloud-based capabilities enable you to:
- Monitor the status of your instrument from your PC or Mac® computer
- Monitor the status of your instrument from your mobile device with the Instrument Connect app
- Upload measured data automatically or manually from your instrument to your cloud account
- Securely store, access, share, and manage your data remotely
- Receive automatic notification of new instrument software updates
- Upgrade instrument software at your convenience
Documents
Specifications
- Description:Multiskan SkyHigh with touch screen and µDrop Duo
- Incubation range:From ambient temperature to 45 ˚C
- Light source:Xenon flash lamp
- Shaking:Linear
- Measurement time (96-well plate):6 s
- Measurement time (384-well plate):10 s
- Range:Up to 4 Abs
- Scan speed:10 s from 200 - 1000 nm in 1 nm steps
- Weight:11,3 kg
- Interfaces:1× USB B port for PC
1× Ethernet port
3× USB A ports for devices (USB memory device and Wi-Fi dongle) - Cat. No.:736-0931
- Accuracy:1,0% + 0,003 Abs (0 - 2,0 Abs), 2,0% (2,0 - 2,5 Abs)
- Wavelength selection:Monochromator
- Precision:SD <0,003 Abs or CV <1,0% (at 450 nm)
- User interface:Stand alone use: 7" touch screen display; PC control: Thermo Scientific™ SkanIt™ software
- Linearity:0 - 2,5 Abs, 2% (at 450 nm)
- Bandwidth:<2,5 nm
- Wavelength range:200 - 1000 nm (1 nm steps)
- W×D×H:295×445×265 mm
- Plate types:6- to 48-, 96-, and 384-well plates (max. height 19,5 mm with lid), Thermo Scientific™ µDrop™ Plates
- Cuvette type:W×D×H: 12,5×12,5×40 - 58 mm
Beam centre height: 8,5 mm
Beam window: ≥2
Accessories
Product Information
- Catalog #AvailabilityPrice
- µDrop™ Plate, 16 measurement positions in a 2×8 matrixµDrop™ Plate, 16 measurement positions in a 2×8 matrix736-0754Log in for availability1 * 1 UN - 3 290,00 €
Product Information
- Catalog #AvailabilityPrice
- µDrop™ Duo plate, 2× 16 measurement positions in two 2×8 matrixesµDrop™ Duo plate, 2× 16 measurement positions in two 2×8 matrixesLABLN12391M2Log in for availability1 * 1 UN - 3 570,00 €
Las recomendaciones se personalizarán según sus preferencias de compra solo si ha dado su consentimiento activando la opción "Mejorar mi experiencia de compra" en la "página de información personal".
De lo contrario, recibirá recomendaciones genéricas.



