Order Entry
Spain
Orders LinkContactUsLinkComponent
Microplate readers, Multiskan SkyHigh

Microplate readers, Multiskan SkyHigh

Multiskan SkyHigh
Catalog # 736-0923
Microplate readers, Multiskan SkyHigh
Catalog # 736-0923
Microplate readers, Multiskan SkyHigh
Microplate readers, Multiskan SkyHigh

Multiskan SkyHigh

Price
15 160,00 €
Log in for availability
1 * 1 UN
Product Details & Documents

The Multiskan SkyHigh is a UV/Visible microplate spectrophotometer designed to be convenient and easy to use for virtually any photometric research application, especially DNA, RNA and protein analysis. It is ideal for multi-user environments where a variety of endpoint, kinetic and spectral assays are performed.

  • Fast, easy measurement - low volume nucleic acid or protein samples
  • Simple to use software - enables experiment set-up within minutes
  • Remote access - manage your data from anywhere via the Cloud, USB, or the Microsoft™ OneDrive™ platform

Multiskan Sky touch screen models can be connected to Thermo Fisher Connect cloud-based tools to enable secure digital data management, remote access of your instrument’s photometric data, and sharing between colleagues.

SkanIt™ software offers a comprehensive selection of built-in calculations, including blank subtraction, standard curves and concentration/dose-response curve fitting, classification, quality control, kinetic, and spectral analysis. Each user can install SkanIt software on his/her own computer to set up assays and analyse data. Thermo Fisher Connect cloud-based capabilities enable you to:
- Monitor the status of your instrument from your PC or Mac® computer
- Monitor the status of your instrument from your mobile device with the Instrument Connect app
- Upload measured data automatically or manually from your instrument to your cloud account
- Securely store, access, share, and manage your data remotely
- Receive automatic notification of new instrument software updates
- Upgrade instrument software at your convenience

Specifications
  • Description:
    Multiskan SkyHigh
  • Incubation range:
    From ambient temperature to 45 ˚C
  • Light source:
    Xenon flash lamp
  • Shaking:
    Linear
  • Measurement time (96-well plate):
    6 s
  • Measurement time (384-well plate):
    10 s
  • Range:
    Up to 4 Abs
  • Scan speed:
    10 s from 200 - 1000 nm in 1 nm steps
  • Weight:
    11,3 kg
  • Interfaces:
    1× USB B port for PC
    1× Ethernet port
    3× USB A ports for devices (USB memory device and Wi-Fi dongle)
  • Cat. No.:
    736-0923
  • Accuracy:
    1,0% + 0,003 Abs (0 - 2,0 Abs), 2,0% (2,0 - 2,5 Abs)
  • Wavelength selection:
    Monochromator
  • Precision:
    SD <0,003 Abs or CV <1,0% (at 450 nm)
  • User interface:
    Stand alone use: 7" touch screen display; PC control: Thermo Scientific™ SkanIt™ software
  • Linearity:
    0 - 2,5 Abs, 2% (at 450 nm)
  • Bandwidth:
    <2,5 nm
  • Wavelength range:
    200 - 1000 nm (1 nm steps)
  • W×D×H:
    295×445×265 mm
  • Plate types:
    6- to 48-, 96-, and 384-well plates (max. height 19,5 mm with lid), Thermo Scientific™ µDrop™ Plates
  • Cuvette type:
    W×D×H: 12,5×12,5×40 - 58 mm
    Beam centre height: 8,5 mm
    Beam window: ≥2
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • µDrop™ Plate, 16 measurement positions in a 2×8 matrix
    µDrop™ Plate, 16 measurement positions in a 2×8 matrix
    736-0754
    Log in for availability
    1 * 1 UN - 3 290,00 €

Product Information

  • Catalog #
    Availability
    Price
  • µDrop™ Duo plate, 2× 16 measurement positions in two 2×8 matrixes
    µDrop™ Duo plate, 2× 16 measurement positions in two 2×8 matrixes
    LABLN12391M2
    Log in for availability
    1 * 1 UN - 3 570,00 €