Order Entry
Spain
ContactUsLinkComponent
High-precision temperature measurement: The digital penetration probe (with the compatible measuring instrument) takes measurements in liquids and pastes, with an accuracy of up to ±0.15 °C. Accuracy of up to ±0.15 °C For measurements in liquids and 1 * 1 items
Catalog # TEST0618.0073
Supplier:  Testo
High-precision temperature measurement: The digital penetration probe (with the compatible measuring instrument) takes measurements in liquids and pastes, with an accuracy of up to ±0.15 °C. Accuracy of up to ±0.15 °C For measurements in liquids and 1 * 1 items
Catalog # TEST0618.0073
Supplier:  Testo
Frequently Bought Together