Order Entry
Spain
Orders LinkContactUsLinkComponent
Re-Usable Cleaning Coupons
Re-Usable Cleaning Coupons
304 stainless steel - bright, 50×50 mm
Catalog # 300-0610
Supplier:   Sampling Systems
Re-Usable Cleaning Coupons
Catalog # 300-0610
Supplier:  Sampling Systems
Re-Usable Cleaning Coupons
Re-Usable Cleaning Coupons
304 stainless steel - bright, 50×50 mm
Price
272,00 €
Log in for availability
1 * 1 UN
Product Details & Documents
Cleaning coupons are great for validating a cleaning regime.
  • All popular sizes available
  • Wide variety of materials and finishes
Sampling Systems' extensive range of cleaning coupons come in a diverse selection of materials, sizes, and finishes. They are ideal for validating cleaning regimes. Popular materials include 316 stainless steel, PTFE, aluminium, and HDPE.
Specifications
  • Description:
    304 stainless steel - bright
  • L×W:
    50×50 mm
  • Seq:
    02