Order Entry
Spain
ContactUsLinkComponent
medical mask op air-10, tie on EN 14683 type II, blue 1 * 300 items
Catalog # HOPEM21101-30
Supplier:  Medicom
undefined
medical mask op air-10, tie on EN 14683 type II, blue 1 * 300 items
Catalog # HOPEM21101-30
Supplier:  Medicom