Order Entry
Spain
ContactUsLinkComponent
15 MM BORE X 150 MM LENGTH FIXED/ADJUSTABLE OMNIFIT COLUMN 1 * 1 items
  MSPP30-3301
15 MM BORE X 150 MM LENGTH FIXED/ADJUSTABLE OMNIFIT COLUMN 1 * 1 items
  MSPP30-3301
Frequently Bought Together