MP Knee-High Boot, Pair of Two, 45.7 cm Tall, 35.6 cm Opening Size, 2XL/3XL, Sterile 1 * 100 items
Catalog # VELT1700PMCNS17041
Supplier: VELTEK ASSOCIATES
MP Knee-High Boot, Pair of Two, 45.7 cm Tall, 35.6 cm Opening Size, 2XL/3XL, Sterile 1 * 100 items
Catalog # VELT1700PMCNS17041
Supplier: VELTEK ASSOCIATES
Frequently Bought Together
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection