Heating Mantles WHM, for beakers 10.000 ml,without integrated power controller, temp. range 450°C,1400 W, power supply 230 V 1 * 1 items
WITGDH.WHM121791
Heating Mantles WHM, for beakers 10.000 ml,without integrated power controller, temp. range 450°C,1400 W, power supply 230 V 1 * 1 items
WITGDH.WHM121791
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
VWR® Facial Tissues
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK