Order Entry
Spain
ContactUsLinkComponent
SEA STAR VORTEXER SPEED RANGE: VARIABE 500-3000RPM (SHAKING OR VORTEXING) 3 POSITION SWITCH ORBIT: 4.2MM 90-110V AC, 50/60 HZ 1 * 1 UN
Catalog # MSPP6100-800
SEA STAR VORTEXER SPEED RANGE: VARIABE 500-3000RPM (SHAKING OR VORTEXING) 3 POSITION SWITCH ORBIT: 4.2MM 90-110V AC, 50/60 HZ 1 * 1 UN
Catalog # MSPP6100-800
Frequently Bought Together