Order Entry
Spain
ContactUsLinkComponent
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects Emmprin in Huma n samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC. 1 * 1 KIT
  BSBTEZ0751
A ELISA kit containing the core components for developing solid phase sandwich ELISA assay to quantitatively detects Emmprin in Huma n samples. The kit contains antibody pairs, recombinant protein standards and HRP conjugated ABC. 1 * 1 KIT
  BSBTEZ0751
Frequently Bought Together