Order Entry
Spain
ContactUsLinkComponent
Sandwich High Sensitivity ELISA kit for Quantitative Detection of rat FGF23. 96wells/kit, with removable strips. 1 * 1 KIT
Catalog # BSBTEK1591
Supplier:  Boster Bio
Sandwich High Sensitivity ELISA kit for Quantitative Detection of rat FGF23. 96wells/kit, with removable strips. 1 * 1 KIT
Catalog # BSBTEK1591
Supplier:  Boster Bio
Frequently Bought Together