Re-Usable Cleaning Coupons
Stainless steel 316L - satin, 50×50 mm
Catalog # SAMP8644S6-50S
Supplier: Sampling Systems
Jump to:
Product Details & Documents
Cleaning coupons are great for validating a cleaning regime.
- All popular sizes available
- Wide variety of materials and finishes
Sampling Systems' extensive range of cleaning coupons come in a diverse selection of materials, sizes, and finishes. They are ideal for validating cleaning regimes. Popular materials include 316 stainless steel, PTFE, aluminium, and HDPE.
Specifications
- Description:Stainless steel 316L - satin
- L×W:50×50 mm
- Seq:04
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



