Order Entry
Spain
Orders LinkContactUsLinkComponent
Bendamustine Related Compound, Reference Standard

Bendamustine Related Compound, Reference Standard

Bendamustine Related Compound D, USP reference standard, Vial, 20 mg
Catalog # 1065265.
Supplier:   USP
Bendamustine Related Compound, Reference Standard
Catalog # 1065265.
Supplier:  USP
CAS Number:  
Bendamustine Related Compound, Reference Standard

Bendamustine Related Compound D, USP reference standard, Vial, 20 mg

CAS Number:  
Price
1 210,00 €
Log in for availability
1 * 20 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    20 mg
  • Standards:
    USP reference standard
  • Description:
    Bendamustine Related Compound D
  • Cat. No.:
    1065265.
  • Formula:
    C₁₄H₁₉Cl₂N₃O₂
  • Description:
    Bendamustine Related Compound D
  • Synonym(s):
    4-{5-[(2-Chloroethyl)amino]-1-methyl-1H -benzimidazol-2-yl}butanoic acid hydrochloride
  • CAS No.:
    1797881-48-1

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.