Order Entry
Spain
ContactUsLinkComponent
Dispenser Ceramus 1-5MlPiston made of high-purity aluminium oxide ceramic, Threaded suction hose fitting, steam sterilised at 121° C without dismantling 1 * 1 items
Catalog # HIRS9332001
Supplier:  HIRSCHMANN
Dispenser Ceramus 1-5MlPiston made of high-purity aluminium oxide ceramic, Threaded suction hose fitting, steam sterilised at 121° C without dismantling 1 * 1 items
Catalog # HIRS9332001
Supplier:  HIRSCHMANN
Frequently Bought Together