Order Entry
Spain
ContactUsLinkComponent
GDF2 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human GDF2 in bone tissue, cell culture supernates, serum, plasma (heparin, EDTA) and urine. 1 * 1 KIT
Catalog # ABNOKA5188
Supplier:  Abnova
GDF2 (Human) ELISA Kit is a sandwich enzyme-linked immunosorbent assay for quantitative detection of human GDF2 in bone tissue, cell culture supernates, serum, plasma (heparin, EDTA) and urine. 1 * 1 KIT
Catalog # ABNOKA5188
Supplier:  Abnova
Frequently Bought Together