Order Entry
Spain
ContactUsLinkComponent
Stericlin® barcode-labels, on roll, for thermal transfer printers, paper, single self-adhesive, with STEAM indicator, roll width 70m m x length 80m x core 50mm 1 * 5 Roll
Catalog # VEPA3FSKS675108
Stericlin® barcode-labels, on roll, for thermal transfer printers, paper, single self-adhesive, with STEAM indicator, roll width 70m m x length 80m x core 50mm 1 * 5 Roll
Catalog # VEPA3FSKS675108
Frequently Bought Together