Order Entry
Spain
ContactUsLinkComponent
Stericlin® tray liner, made of white crepe paper, to protect wrapping materials from mesh trays, dimensions 35x35cm, pre-formed for tray size 25x25cm 1 * 500 items
Catalog # VEPA3FTSZ420102
Stericlin® tray liner, made of white crepe paper, to protect wrapping materials from mesh trays, dimensions 35x35cm, pre-formed for tray size 25x25cm 1 * 500 items
Catalog # VEPA3FTSZ420102
Frequently Bought Together