Order Entry
Spain
Orders LinkContactUsLinkComponent
SILVER NITRATE STD SOLUTION 0.100N 1 * 1 L

SILVER NITRATE STD SOLUTION 0.100N 1 * 1 L

Catalog # 705-1123
Supplier:   Hach
SILVER NITRATE STD SOLUTION 0.100N 1 * 1 L
Catalog # 705-1123
Supplier:  Hach
CAS Number:  
SILVER NITRATE STD SOLUTION 0.100N 1 * 1 L
CAS Number:  
Price
63,40 €
Log in for availability
1 * 1 L
Product Details & Documents