Agilent J+W DB-5HT Intuvo GC column module, 15 m, 0.25 mm, 0.10 µm 1 * 1 items
Catalog # HEWL122-5711-INT
Proveedor: Agilent
Agilent J+W DB-5HT Intuvo GC column module, 15 m, 0.25 mm, 0.10 µm 1 * 1 items
Catalog # HEWL122-5711-INT
Proveedor: Agilent
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Proveedor: HEATHROW
Acetone ≥99%, TECHNICAL
Proveedor: VWR Chemicals
VWR® Facial Tissues
Proveedor: VWR Collection
VWR®, Nitrile Gloves
Proveedor: VWR Collection
pH indicator strips, MQuant®, Supelco®
Proveedor: Merck
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Proveedor: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Proveedor: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Proveedor: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Proveedor: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Proveedor: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Proveedor: VWR Collection
VWR®, Transfer Pipettes, High Performance
Proveedor: VWR Collection