Order Entry
Spain
ContactUsLinkComponent
Agilent J+W DB-225ms Intuvo GC column module 30 m, 0.25 mm, 0.25 µm 1 * 1 items
Catalog # HEWL122-2932-INT
Supplier:  Agilent
Agilent J+W DB-225ms Intuvo GC column module 30 m, 0.25 mm, 0.25 µm 1 * 1 items
Catalog # HEWL122-2932-INT
Supplier:  Agilent
Frequently Bought Together