Entrada de pedido
Spain
ContactUsLinkComponent
[EN]DB-HEAVYWAX 30M, 0.25MM, 0.25UM 1 * 1 UN
Catalog # HEWL122-7132
Supplier:  Agilent
undefined
[EN]DB-HEAVYWAX 30M, 0.25MM, 0.25UM 1 * 1 UN
Catalog # HEWL122-7132
Supplier:  Agilent
Frequently Bought Together