Order Entry
Spain
Orders LinkContactUsLinkComponent
Safety spectacles, Solus™ 1000
Anti-UV (2C-1.2), anti-scratch, anti-fog, Green/black, Amber PC
  111-2378
 :   3M
Safety spectacles, Solus™ 1000
  111-2378
 :  3M
Safety spectacles, Solus™ 1000
Safety spectacles, Solus™ 1000
Anti-UV (2C-1.2), anti-scratch, anti-fog, Green/black, Amber PC
23,20 €
1 * 1 UN
3M™ Solus™ 1000 series safety glasses feature a strong polycarbonate lens in a slim, stylish frame. These glasses feature a Scotchgard™ Anti-fog/anti-scratch coating, and a soft temple and nose bridge.
  • Stylish and modern slim frame with colour accented soft temple components
  • Strong polycarbonate lens for impact resistance
  • Scotchgard™ Anti-Fog coating offers superior anti-fog and anti-scratch properties, even after washing multiple times
  • Lightweight with soft cushioning on nose bridge
  • Suitable for automatic cleaning, disinfection, and sterilisation by autoclaving at 121 °C
  • Inner padding on temples for increased comfort and stability
  • Excellent protection against ultraviolet (UV) radiation
Optional: Adjustable headband and foam or TPE gasket.
 : EN 166
  • Type:
    Anti-UV (2C-1.2), anti-scratch, anti-fog
  • Colour:
    Green/black
  • Seq:
    06
  • Lenses:
    Amber PC
Accessories
  •  
    Description
  • MMMASOLUSSR
    Foam insert for 3M™ Solus™ 1000 spectacles, for a closer fit to the face
    1 * 5 UN - 39,90 €