Order Entry
Spain
ContactUsLinkComponent
Storage Boxes for CryoSette Frozen Tissue containers. 21 palces. Made of polypropylene. Compatible with leading frozen sample stora ge systems. Can be handled by robotic equipment. Autoclavable. Temperature range -196 C to 110 C. Dimensions: 127.7 1 * 1 items
Catalog # SIMPM956-21W
Supplier:  Simport Scientific
Storage Boxes for CryoSette Frozen Tissue containers. 21 palces. Made of polypropylene. Compatible with leading frozen sample stora ge systems. Can be handled by robotic equipment. Autoclavable. Temperature range -196 C to 110 C. Dimensions: 127.7 1 * 1 items
Catalog # SIMPM956-21W
Supplier:  Simport Scientific
Frequently Bought Together