Order Entry
Spain
ContactUsLinkComponent
Disposable Tip (DiTi) Box, empty for use with 10/50/200 µL refill insert tips, ANSI Format. Tecan® PURE purity level - tested and c ertified to be free of human DNA, Rnase, Dnase and PCR inhibitors. For use with the Freedom EVO® Liquid Handling Arm™ 1 * 10 items
Catalog # TCAN30058506
Supplier:  Tecan
Disposable Tip (DiTi) Box, empty for use with 10/50/200 µL refill insert tips, ANSI Format. Tecan® PURE purity level - tested and c ertified to be free of human DNA, Rnase, Dnase and PCR inhibitors. For use with the Freedom EVO® Liquid Handling Arm™ 1 * 10 items
Catalog # TCAN30058506
Supplier:  Tecan
Frequently Bought Together