Order Entry
Spain
ContactUsLinkComponent
The HIGH FLOW TETPOR II is validated as a 0.2 micron sterilizing grade filter in liquids through ASTM F838-05 and 0.01 micron in gas through full retention to an aerosol challenge of MS2 phage. 1 * 3 items
Catalog # PARKZHFT/BT
Supplier:  PARKER HANNIFIN
The HIGH FLOW TETPOR II is validated as a 0.2 micron sterilizing grade filter in liquids through ASTM F838-05 and 0.01 micron in gas through full retention to an aerosol challenge of MS2 phage. 1 * 3 items
Catalog # PARKZHFT/BT
Supplier:  PARKER HANNIFIN
Frequently Bought Together