Order Entry
Spain
Orders LinkContactUsLinkComponent
VISOCOLOR® Colorimetric Test, Hydrazine

VISOCOLOR® Colorimetric Test, Hydrazine

VISOCOLOR® ECO Hydrazine, test no. 5-30, Reagents for 130 tests, color chart and accessories in a box, 0 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 • 0,40 mg/L (N₂H₄); 0,05 - 0,40 mg/L (N₂H₄), 4-Dimethylaminobenzaldehyde, PF-12Plus;Advance;Visual, 168×112×40 mm (Including packaging), 130, 1
Catalog # MANA931030
Supplier:   MACHEREY-NAGEL
VISOCOLOR® Colorimetric Test, Hydrazine
Catalog # MANA931030
Supplier:  MACHEREY-NAGEL
CAS Number:  
VISOCOLOR® Colorimetric Test, Hydrazine
VISOCOLOR® Colorimetric Test, Hydrazine

VISOCOLOR® ECO Hydrazine, test no. 5-30, Reagents for 130 tests, color chart and accessories in a box, 0 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 • 0,40 mg/L (N₂H₄); 0,05 - 0,40 mg/L (N₂H₄), 4-Dimethylaminobenzaldehyde, PF-12Plus;Advance;Visual, 168×112×40 mm (Including packaging), 130, 1

CAS Number:  
Price
98,50 €
Log in for availability
1 * 1 UN
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Reagents for the determination of hydrazine in water samples.

  • Combines simple implementation with high measurement reliability

Simple determination directly on site or in the laboratory.

Sea water analysis: Yes

Hazardous material: Yes

Remark: Measuring range on PF-12Plus. Range on other photometers can be different.

Specifications

Specifications

  • Chemical method:
    4-Dimethylaminobenzaldehyde
  • Pk:
    1
  • Description:
    VISOCOLOR® ECO Hydrazine, test no. 5-30
  • Dimensions:
    168×112×40 mm (Including packaging)
  • For unit:
    PF-12Plus;Advance;Visual
  • Tests per kit:
    130
  • Range:
    0 • 0,05 • 0,10 • 0,15 • 0,20 • 0,25 • 0,30 • 0,40 mg/L (N₂H₄); 0,05 - 0,40 mg/L (N₂H₄)
  • Contents:
    Reagents for 130 tests, color chart and accessories in a box
  • Cat. No.:
    MANA931030
  • Platform:
    Colorimetric test kits
  • Model:
    MANA931030
  • Shelf life (months):
    12
  • Storage temperature:
    15...25 °C
  • Weight:
    237 g (Including packaging)

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.